Cholecystokinin, CCK Octapeptide (26-33)
Cholecystokinin, CCK Octapeptide (26-33)
Cholecystokinin, CCK Octapeptide (26-33)
Thermo Scientific Chemicals

Cholecystokinin, CCK Octapeptide (26-33)

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66669.EXD
also known as J66669-!D
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS25126-32-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Suppliers CofAPeptide Purity: ≥95.0%
FormLyophilized Powder
Mass SpecExhibits correct MW
Cholecystokinin, CCK Octapeptide (26-33) is used in the biological studies in the preparation of opioid receptors agonistic and CCK receptor antagonistic activity. It plays a role in satiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C