Chemical Identifiers
CAS34642-77-8
IUPAC Namesodium (2S,5R,6R)-6-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC16H18N3NaO5S
InChI KeyBYHDFCISJXIVBV-YWUHCJSESA-M
SMILES[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=C(O)C=C3)C(=O)N2[C@H]1C([O-])=O
View more