1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%
Thermo Scientific Chemicals

1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%

CAS: 4163-65-9 | C16H22O11 | 390.341 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J66685.14
also known as J66685-14
Price (USD)
296.65
Online Exclusive
329.00
Save 32.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
296.65
Online Exclusive
329.00
Save 32.35 (10%)
Each
Add to cart
Specifications
CAS4163-65-9
Chemical Name or Material1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose
ColorWhite
Formula Weight390.34
IUPAC Name[(2R,3R,4S,5S,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
View more
α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only