Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles
Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles
Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles
Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles
Thermo Scientific Chemicals

Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles

20-50nm particles
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
250 mL
Catalog number J66691.AK
also known as J66691-AK
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS12167-74-7
IUPAC Nametricalcium diphosphate
Molecular FormulaCa3O8P2
InChI KeyQORWJWZARLRLPR-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Particle size: 20-50nm
Assay (unspecified)Percent solids: 5-10%
Hydroxyapatite nanoparticles is used for polishing and dental powders, porcelain and pottery, and enameling. It is also utilized to engineer new biomaterials for applications such as bone grafts and fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C