Cefamandole nafate
Cefamandole nafate
Cefamandole nafate
Thermo Scientific Chemicals

Cefamandole nafate

A second generation cephalosporin antibiotic | CAS: 42540-40-9 | C19H17N6NaO6S2 | 512.491 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66735.06
also known as J66735-06
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Specifications
CAS42540-40-9
ChEBICHEBI:3481
Chemical Name or MaterialCefamandole nafate
EINECS Number255-877-4
Formula Weight513.5
View more
A second generation cephalosporin antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only