Teicoplanin, cont. 80+% A2
Teicoplanin, cont. 80+% A2
Teicoplanin, cont. 80+% A2
Thermo Scientific Chemicals

Teicoplanin, cont. 80+% A2

CAS: 61036-62-21879.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100mg
500mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66785.ME
also known as J66785-ME
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Quantity:
500mg
Request bulk or custom format
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS61036-62-2
IUPAC Name(1S,2R,19R,22R,34S,37R,40R,52R)-22-amino-63,65-dichloro-64-{[(2S,3R,4R,5S,6R)-3-decanamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-26,31,44,49-tetrahydroxy-21,35,38,54,56,59-hexaoxo-47-{[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-7,13,28-trioxa-20,36,39,53,55,58-hexaazaundecacyclo[38.14.2.2³,⁶.2¹⁴,¹⁷.2¹⁹,³⁴.1⁸,¹².1²³,²⁷.1²⁹,³³.1⁴¹,⁴⁵.0¹⁰,³⁷.0⁴⁶,⁵¹]hexahexaconta-3,5,8,10,12(64),14,16,23,25,27(61),29,31,33(60),41,43,45(57),46,48,50,62,65-henicosaene-52-carboxylic acid
Molecular FormulaC88H97Cl2N9O33
InChI KeyBJNLLBUOHPVGFT-QRZIFLFXSA-N
SMILESCCCCCCCCCC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=C2OC3=CC=C(C[C@H]4NC(=O)[C@H](N)C5=CC=C(O)C(OC6=CC(O)=CC(=C6)[C@H](NC4=O)C(=O)N[C@@H]4C(C=C1OC1=CC=C(C=C1Cl)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O)[C@@H]1NC(=O)[C@H](NC4=O)C4=CC=C(O)C(=C4)C4=C(O[C@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]6O)C=C(O)C=C4[C@@H](NC1=O)C(O)=O)=C2)=C5)C=C3Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥900 IU/mg (on dried basis)
Assay from Supplier's CofAA2 Group: ≥80.0%
Assay from Supplier's CofAA2-2: 35.0-55.0%
Assay from Supplier's CofAA2-3: ≤20.0%
Assay from Supplier's CofAA2-4: ≤20.0%
View more

A glycopeptide antibiotic produced by Actinoplanes teichomiceticus; effective against Gram-positive bacteria that are resistant to β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
Keep cold