Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals

A broad spectrum antibiotic
Have Questions?
Catalog NumberQuantity
J66800.03
also known as J66800-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66800.03
also known as J66800-03
Price (USD)
180.65
Online exclusive
201.00
Save 20.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
180.65
Online exclusive
201.00
Save 20.35
Each
Chemical Identifiers
CAS33564-30-6
IUPAC Namesodium (6R,7S)-3-[(carbamoyloxy)methyl]-7-methoxy-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular FormulaC16H16N3NaO7S2
InChI KeyGNWUOVJNSFPWDD-XMZRARIVSA-M
SMILES[Na+].CO[C@]1(NC(=O)CC2=CC=CS2)[C@H]2SCC(COC(N)=O)=C(N2C1=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormWhite solid
Assay≥ 97%
Assay (unspecified)≥ 97%
A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only