1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
Thermo Scientific Chemicals

1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose

CAS: 97614-43-2 | C26H21FO7 | 464.445 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66805.06
also known as J66805-06
Price (USD)
172.65
Online Exclusive
192.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
172.65
Online Exclusive
192.00
Save 19.35 (10%)
Each
Add to cart
Specifications
CAS97614-43-2
Chemical Name or Material1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
ColorWhite
Formula Weight464.44
IUPAC Name[(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate
View more
It is a pharmaceutical intermediate and is used as an OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used as an OLED materials.

Solubility
Soluble in chloroform or dichloromethane

Notes
Refrigerate. Keep away from heat, oxidizing agents.
RUO – Research Use Only