[Val<sup>5</sup>] Angiotensin II, Human
Thermo Scientific™

[Val5] Angiotensin II, Human

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66806.LB0
also known as J66806-LB0
5 mg
Catalog number J66806.LB0
also known as J66806-LB0
Price (USD)
97.65
Special offer
Online exclusive
Ends: 28-Dec-2026
115.00
Save 17.35
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
97.65
Special offer
Online exclusive
Ends: 28-Dec-2026
115.00
Save 17.35
Each
Specifications
FormatSolid
Molecular Weight (g/mol)1032.15
Product Type[Val5] Angiotensin II, human
Purification MethodHPLC
Quantity5 mg
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For Research Use Only
Contents & Storage
-30°C to -10°C