Pyrazinamide, 98%
Pyrazinamide, 98%
Pyrazinamide, 98%
Thermo Scientific Chemicals

Pyrazinamide, 98%

CAS: 98-96-4 | C5H5N3O | 123.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66822.09
also known as J66822-09
Price (USD)
42.65
Online exclusive
47.70
Save 5.05
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
42.65
Online exclusive
47.70
Save 5.05
Each
Specifications
Beilstein112306
CAS98-96-4
ChEBICHEBI:45285
Chemical Name or MaterialPyrazinamide
EINECS Number202-717-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only