Methyl beta-D-ribofuranoside, 98%
Methyl beta-D-ribofuranoside, 98%
Methyl beta-D-ribofuranoside, 98%
Methyl beta-D-ribofuranoside, 98%
Thermo Scientific Chemicals

Methyl beta-D-ribofuranoside, 98%

CAS: 7473-45-2 | C6H12O5 | 164.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J66828.14
also known as J66828-14
Price (USD)
267.65
Special offer
Online exclusive
Ends: 30-Jun-2026
315.00
Save 47.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
267.65
Special offer
Online exclusive
Ends: 30-Jun-2026
315.00
Save 47.35 (15%)
Each
Add to cart
Specifications
CAS7473-45-2
Chemical Name or MaterialMethyl beta-D-ribofuranoside
ColorWhite
EINECS Number231-271-5
Formula Weight164.16
View more
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography. 

Solubility
Soluble in water

Notes
Store at 2-8°C. Incompatible with heat, oxidizing agents.
RUO – Research Use Only

General References:

  1. J. Raap.; J. H. Van Boom.; H. C. Van Lieshout.; C. A. G. Haasnoot. Conformations of methyl 2'-deoxy-.alpha.-D-ribofuranoside and methyl 2'-deoxy-.beta.-D-ribofuranoside. A proton magnetic resonance spectroscopy and molecular mechanics study.J. Am. Chem. Soc. 1988, 110 (9),2736-2743 .