Corticotropin-Releasing Factor, human, rat
Thermo Scientific Chemicals

Corticotropin-Releasing Factor, human, rat

 
Have Questions?
Catalog NumberQuantity
J66835.EXD
also known as J66835-!D
0.5mg
Catalog number J66835.EXD
also known as J66835-!D
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Quantity:
0.5mg
Request bulk or custom format
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS86784-80-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
AssayPeptide content: report value
AssayPurity: >90%
Assay (unspecified)Peptide content: report value
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only