2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
Thermo Scientific Chemicals

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

CAS: 60933-68-8 | C26H28O5 | 420.505 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number J66853.09
also known as J66853-09
Price (USD)
254.65
Online Exclusive
283.00
Save 28.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
254.65
Online Exclusive
283.00
Save 28.35 (10%)
Each
Add to cart
Specifications
CAS60933-68-8
Chemical Name or Material2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
ColorWhite
Formula Weight420.5
IUPAC Name(2R,3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
View more
An arabinofuranose derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An arabinofuranose derivative.

Solubility
Soluble in methanol

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only