PACAP (6-27), Human, Ovine, Rat
Thermo Scientific Chemicals

PACAP (6-27), Human, Ovine, Rat

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66872.EXD
also known as J66872-!D
Price (USD)
166.00
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
166.00
Each
Chemical Identifiers
CAS136134-68-4
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized powder
Assay from Supplier's CofAPeptide purity: ≥90.0% (HPLC)
Assay from Supplier's CofAPeptide content: ≥70.0%
Appearance (Color)White
Mass SpecExhibits correct MW (2638.1 g/mol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C