This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66890.MC
also known as J66890-MC
Price (USD)
667.65
Online Exclusive
742.00
Save 74.35 (10%)
Each
Quantity:
100mg
Request bulk or custom format
Price (USD)
667.65
Online Exclusive
742.00
Save 74.35 (10%)
Each
Specifications
Chemical Name or MaterialMupirocin lithium salt
CAS73346-79-9
Recommended StorageKeep cold
TSCANo
Molecular FormulaC26H43LiO9
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.