Fibronectin-Binding Protein
Thermo Scientific Chemicals

Fibronectin-Binding Protein

For research use
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5mg
1mg
Catalog number J66903.EXD
also known as J66903-!D
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Quantity:
0.5mg
Request bulk or custom format
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS119977-20-7
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAPeptide purity: ≥94.0% (HPLC)
Appearance (Color)White
FormLyophilized powder
Assay from Supplier's CofAPeptide Content: Report %
Mass SpecElectrospray exhibits correct molecular weight
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only