3,4,6-Tri-O-acetyl-D-galactal
3,4,6-Tri-O-acetyl-D-galactal
3,4,6-Tri-O-acetyl-D-galactal
Thermo Scientific Chemicals

3,4,6-Tri-O-acetyl-D-galactal

CAS: 4098-06-0 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number J66904.03
also known as J66904-03
Price (USD)
43.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.30
Each
Specifications
CAS4098-06-0
Chemical Name or Material3,4,6-Tri-O-acetyl-D-galactal
ColorYellow
EINECS Number223-859-5
Flash Point>110°C (235°F)
View more
3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

Solubility
Soluble in chloroform at 10mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Jean-Claude Jacquinet. Syntheses of the methyl glycosides of the repeating units of chondroitin 4- and 6-sulfate. Carbohydrate Research. 1990, 199, (2), 153-181.
  2. Francesco Gonzalez.; Suzanne Lesage.; Arthur S. Perlin. Catalysis by mercuric ion of reactions of glycals with water ☆. Carbohydrate Research. 1975, 42, (2), 267-274.