3,4-Di-O-acetyl-6-deoxy-L-glucal, 98%
3,4-Di-O-acetyl-6-deoxy-L-glucal, 98%
3,4-Di-O-acetyl-6-deoxy-L-glucal, 98%
3,4-Di-O-acetyl-6-deoxy-L-glucal, 98%
Thermo Scientific Chemicals

3,4-Di-O-acetyl-6-deoxy-L-glucal, 98%

CAS: 34819-86-8 | C10H14O5 | 214.217 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66944.06
also known as J66944-06
Price (USD)
185.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
185.00
Each
Add to cart
Chemical Identifiers
CAS34819-86-8
IUPAC Name3-(acetyloxy)-2-methyl-3,4-dihydro-2H-pyran-4-yl acetate
Molecular FormulaC10H14O5
InChI KeyNDEGMKQAZZBNBB-UHFFFAOYNA-N
SMILESCC1OC=CC(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Assay from Supplier's CofA≥97.5% (1H-NMR)
Optical Rotation+50° to +60° (c=1 in chloroform @ 20°C)
Appearance (Color)Colorless
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only