Florfenicol, 98%
Florfenicol, 98%
Florfenicol, 98%
Thermo Scientific Chemicals

Florfenicol, 98%

CAS: 73231-34-2 | C12H14Cl2FNO4S | 358.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66995.03
also known as J66995-03
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS73231-34-2
IUPAC Name2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methanesulfonylphenyl)propan-2-yl]acetamide
Molecular FormulaC12H14Cl2FNO4S
InChI KeyAYIRNRDRBQJXIF-NXEZZACHSA-N
SMILESCS(=O)(=O)C1=CC=C(C=C1)[C@@H](O)[C@@H](CF)NC(=O)C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (HPLC)≥98.0%
Melting point150-156°C
A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
RUO – Research Use Only