Silver nanoparticles, 50nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 425nm absorption
Silver nanoparticles, 50nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 425nm absorption
Silver nanoparticles, 50nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 425nm absorption
Thermo Scientific Chemicals

Silver nanoparticles, 50nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 425nm absorption

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number J67065.AC
also known as J67065-AC
Price (USD)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Buffer2 mM sodium citrate
CommentCore size: 50nm ( ± 3nm)
lambda-max (UV-Vis)410-430nm
Particle Size~2.9E+10
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only