Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption
Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption
Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption
Thermo Scientific Chemicals

Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number J67099.AC
also known as J67099-AC
Price (USD)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
AbsorbanceAbsorption (max): 490-515 nm
Assay (unspecified)Concentration (OD): 0.90-0.92
Assay (unspecified)Core Size: 100 nm (-2nm to +15nm)
Assay (unspecified)Hydrodynamic Diameter (z-average): 100-120nm
Assay (unspecified)Particles/mL: ~3.6E+09
An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range
  • Readily binds proteins and other ligands for conjugate and sensor development
RUO – Research Use Only