Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered
Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered
Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals

Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered

Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities. | CAS: 4800-94-6 | C17H16N2Na2O6S | 422.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67159.AD
also known as J67159-AD
Price (USD)
320.65
Online Exclusive
356.00
Save 35.35 (10%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
320.65
Online Exclusive
356.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS4800-94-6
IUPAC Namedisodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC17H16N2Na2O6S
InChI KeyRTYJTGSCYUUYAL-YCAHSCEMSA-L
SMILES[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to light yellow
CommentComponent is dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 0.22 micron filter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium

Application

  • Due to its antibacterial activity this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
RUO – Research Use Only

General References:

  1. Cole, M. R.; Hobden, J. A.; Warner, I. M. Recycling Antibiotics into GUMBOS: A New Combination Strategy to Combat Multi-Drug-Resistant Bacteria. Molecules 2015, 20 (4), 6466-6487.
  2. Sonopo, M. S.; Venter, K.; Boyle, G.; Winks, S.; Painter, B. M.; Zeevaart, J. R. Carbon-14 radiolabeling and in vivo biodistribution of a potential anti-TB compound. J. Labelled Comp. Radiopharm. 2015, 58 (2), 23-29.