Ceftriaxone disodium salt, 50 mg/ml in distilled water, Thermo Scientific Chemicals
Ceftriaxone disodium salt, 50 mg/ml in distilled water, Thermo Scientific Chemicals
Ceftriaxone disodium salt, 50 mg/ml in distilled water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ceftriaxone disodium salt, 50 mg/ml in distilled water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 x 1 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67171.XF
also known as J67171-XF
Price (USD)
98.30
Each
Quantity:
1 mL
Request bulk or custom format
Price (USD)
98.30
Each
Specifications
CAS104376-79-6
Chemical Name or MaterialCeftriaxone disodium salt
EINECS Number277-930-0
Health Hazard 1H317-H334-H335
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only