Boric acid, Electrophoresis Grade,  99.5% min
Boric acid, Electrophoresis Grade,  99.5% min
Boric acid, Electrophoresis Grade,  99.5% min
Thermo Scientific Chemicals

Boric acid, Electrophoresis Grade, 99.5% min

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Catalog number J67202.36
also known as J67202-36
Price (USD)
60.10
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
60.10
Each
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
CommentPhosphate: ≤0.001%
CommentSulfate: ≤0.01%
DNaseNone detected
Heavy Metals≤0.001 % (as Pb)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only