Piperacillin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Piperacillin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Piperacillin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Piperacillin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 x 1 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67261.XF
also known as J67261-XF
Price (USD)
96.50
Each
Quantity:
1 mL
Request bulk or custom format
Price (USD)
96.50
Each
Specifications
CAS59703-84-3
ChEBICHEBI:8233
Chemical Name or MaterialPiperacillin sodium salt
EINECS Number261-868-6
Health Hazard 1H317-H334
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only