N-Arachidonoyl-serotonin, 98%, Thermo Scientific Chemicals
N-Arachidonoyl-serotonin, 98%, Thermo Scientific Chemicals
N-Arachidonoyl-serotonin, 98%, Thermo Scientific Chemicals
N-Arachidonoyl-serotonin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Arachidonoyl-serotonin, 98%, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
J67271.MA
also known as J67271-MA
10 mg
Catalog number J67271.MA
also known as J67271-MA
Price (USD)
133.65
Special offer
Online exclusive
Ends: 30-Jun-2026
157.00
Save 23.35 (15%)
Each
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)
133.65
Special offer
Online exclusive
Ends: 30-Jun-2026
157.00
Save 23.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS187947-37-1
IUPAC NameN-[2-(5-hydroxy-1H-indol-3-yl)ethyl]icosa-5,8,11,14-tetraenamide
Molecular FormulaC30H42N2O2
InChI KeyQJDNHGXNNRLIGA-UHFFFAOYSA-N
SMILESCCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCC1=CNC2=CC=C(O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Iron: 10 ppm max.
Thin Layer Chromatography98.0% min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only