6-Bromoindirubin-3'-oxime, 97%
6-Bromoindirubin-3'-oxime, 97%
6-Bromoindirubin-3'-oxime, 97%
Thermo Scientific Chemicals

6-Bromoindirubin-3'-oxime, 97%

CAS: 667463-62-9 | C16H10BrN3O2 | 356.179 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number J67275.MF
also known as J67275-MF
Price (USD)
691.65
Online Exclusive
768.00
Save 76.35 (10%)
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
691.65
Online Exclusive
768.00
Save 76.35 (10%)
Each
Chemical Identifiers
CAS667463-62-9
IUPAC Name6'-bromo-3-(hydroxyamino)-1',2'-dihydro-[2,3'-biindolyliden]-2'-one
Molecular FormulaC16H10BrN3O2
InChI KeyWNWSUJQVZJJGLF-UHFFFAOYSA-N
SMILESONC1=C2C=CC=CC2=NC1=C1C(=O)NC2=C1C=CC(Br)=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to red to dark red
Assay from Suppliers CofA≥96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only