How should I store reconstituted Interleukin 2, human (Cat. No. J67298.EXE)?
Reconstituted solutions of Interleukin 2, human are stable for up to one week at 2-8°C. For long-term storage of stock solutions, we recommend that you portion the stock into working aliquots and store at -80°C.