Interleukin 2, human - FAQs
Interleukin 2, human - FAQs
View additional product information for Interleukin 2, human - FAQs (J67298.MCR)
2 product FAQs found
How should I store reconstituted Interleukin 2, human (Cat. No. J67298.EXE)?
Reconstituted solutions of Interleukin 2, human are stable for up to one week at 2-8°C. For long-term storage of stock solutions, we recommend that you portion the stock into working aliquots and store at -80°C.
How should I reconstitute Interleukin 2, human (Cat. No. J67298.EXE)?
To reconstitue Interleukin 2, human, we recommend the following:
  • Briefly centrifuge the tube prior to opening it to bring the contents to the bottom
  • Use sterile water to reconstitute to a concentration of 0.1-1.0 mg/mL in a siliconized tube
Further dilutions should be made in appropriate buffered solutions.