Ionomycin calcium salt, 1 mg/ml in methanol, sterile-filtered
Ionomycin calcium salt, 1 mg/ml in methanol, sterile-filtered
Ionomycin calcium salt, 1 mg/ml in methanol, sterile-filtered
Thermo Scientific Chemicals

Ionomycin calcium salt, 1 mg/ml in methanol, sterile-filtered

CAS: 56092-82-1 | C41H70CaO9 | 747.08 g/mol
Have Questions?
Catalog NumberQuantity
J67343.XF
also known as J67343-XF
1 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67343.XF
also known as J67343-XF
Price (USD)
387.65
Online Exclusive
430.00
Save 42.35 (10%)
Each
Quantity:
1 mL
Request bulk or custom format
Price (USD)
387.65
Online Exclusive
430.00
Save 42.35 (10%)
Each
Specifications
CAS56092-82-1
Chemical Name or MaterialIonomycin calcium salt
DOT InformationTransport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
Health Hazard 1H225-H301+H311+H331-H370
Health Hazard 2GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only