Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption
Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption
Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption
Thermo Scientific Chemicals

Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number J67409.AE
also known as J67409-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold nanoparticles
ColorPink to Red
IUPAC Namegold
View more
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
RUO – Research Use Only