HEPES Buffer, 0.2M buffer soln., pH 7.4, low endotoxin
HEPES Buffer, 0.2M buffer soln., pH 7.4, low endotoxin
HEPES Buffer, 0.2M buffer soln., pH 7.4, low endotoxin
Thermo Scientific Chemicals

HEPES Buffer, 0.2M buffer soln., pH 7.4, low endotoxin

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J67485.AK
also known as J67485-AK
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Specifications
CAS7365-45-9
ChEBICHEBI:42334
Chemical Name or MaterialHEPES Buffer
ColorColorless
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only