Product Image
Thermo Scientific Chemicals

HEPES-buffered saline (5X, pH 7.4), low endotoxin

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J67502.AE
also known as J67502-AE
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Specifications
Chemical Name or MaterialHEPES-buffered saline (5X, pH 7.4)
ColorColorless
Name Notelow endotoxin
Recommended StorageKeep cold
Storage Note 12° to 8°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What are the concentrations of the components of the HEPES-buffered saline (5X, pH 7.4), low endotoxin?

The HEPES-buffered saline (5X, pH 7.4), low endotoxin contains 125 mM HEPES and 750 mM Sodium Chloride, prepared in 18.2 megohm-cm±1 low endotoxin water and filtered through a 0.22-micron filter.