Search
View additional product information for Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol - FAQs (J67569.AP, J67569.K2)
1 product FAQs found
The Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol contains: