Krebs-Heneseleit buffer solution - FAQs
Krebs-Heneseleit buffer solution - FAQs
View additional product information for Krebs-Heneseleit buffer solution - FAQs (J67820.AP, J67820.K2)
1 product FAQs found
What are the concentrations of the components of Krebs-Heneseleit buffer solution?
The Krebs-Heneseleit buffer solution contains:
  • 118 mM sodium chloride
  • 4.7 mM potassium chloride
  • 1.25 mM calcium chloride
  • 1.20 mM magnesium sulfate
  • 1.20 mM monopotassium phosphate
  • 25 mM sodium bicarbonate
  • 10 mM HEPES
  • 11.1 mM D-glucose