Search
View additional product information for Krebs-Ringer Solution, without calcium - FAQs (J67839.K2, J67839.AP)
2 product FAQs found
The pH of the Krebs-Ringer Solution, without calcium is 7.3.
The Krebs-Ringer Solution, without calcium contains 120 mM sodium chloride, 5 mM potassium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5.5 mM D-glucose.