Guanidine Hydrochloride, >99.5%, Molecular Biology Grade
Guanidine Hydrochloride, >99.5%, Molecular Biology Grade
Guanidine Hydrochloride, >99.5%, Molecular Biology Grade
Thermo Scientific Chemicals

Guanidine Hydrochloride, >99.5%, Molecular Biology Grade

CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Poly pail
Catalog number J75823.A7
Price (USD)
911.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,072.00
Save 160.35 (15%)
Each
Quantity:
5 kg
Packaging:
Poly pail
Request bulk or custom format
Price (USD)
911.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,072.00
Save 160.35 (15%)
Each
Chemical Identifiers
CAS50-01-1
IUPAC Namehydrogen guanidine chloride
Molecular FormulaCH6ClN3
InChI KeyPJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point184 ? 3?C
Assay (unspecified)Absorbance (225 nm, 6M, 1 cm): ≤ 0.2
Assay (unspecified)Ammeline: None detected
Identification (FTIR)Conforms
Protease activityPasses test
View more
  • Used for extraction of RNA.
  • Preparation of high molecular weight genomic DNA.
  • Solubilizing inclusion bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies
RUO – Research Use Only