TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure
Thermo Scientific Chemicals

TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure

Used in Western blotting protocols | CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
5 L
Packaging:
View all
Nalgene bottle
Catalog number J75892.AE
Price (USD)
66.65
Online Exclusive
74.00
Save 7.35 (10%)
Each
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.00
Save 7.35 (10%)
Each
Specifications
CAS7647-14-5
ChEBICHEBI:26710
Chemical Name or MaterialTBS
IUPAC Namesodium;chloride
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
View more
Used in Western blotting protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What are the concentrations of the components in TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure?

TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure contains 500 mM Tris, 60 mM KCl, and 2.8 M NaCl in high purity RO water.