TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure
Thermo Scientific Chemicals

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure

Use directly to resuspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
10 x 1 mL
500 mL
Packaging:
Nalgene bottle
Catalog number J75893.AE
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.50
Save 12.85 (15%)
Each
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.50
Save 12.85 (15%)
Each
Specifications
CAS1185-53-1
Chemical Name or MaterialTE Buffer
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
Molecular FormulaC4H12ClNO3
View more
Use directly to resuspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What are the concentrations of the components of TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure (Cat. No. J75893.XCR)?

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure consists of 10 mM Tris and 1 mM EDTA in high-purity RO water.