TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure - FAQs
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure - FAQs
View additional product information for TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure - FAQs (J75893.XCR, J75893.AE, J75893.AP)
1 product FAQs found
What are the concentrations of the components of TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure (Cat. No. J75893.XCR)?
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure consists of 10 mM Tris and 1 mM EDTA in high-purity RO water.