TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure - FAQs
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure - FAQs
View additional product information for TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure - FAQs (J75904.K2, J75904.K8)
1 product FAQs found
What are the concentrations of the components of TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure (Cat. No. J75904.K8)?
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure contains 400 mM Tris and 0.01 M EDTA in high-purity RO water.