TBS with Tween™ (TBST), 20X Solution, Molecular Biology Grade, Ultrapure
Thermo Scientific Chemicals

TBS with Tween™ (TBST), 20X Solution, Molecular Biology Grade, Ultrapure

Used in Western blotting protocols
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Catalog number J77500.K8
Price (USD)
402.65
Online Exclusive
447.00
Save 44.35 (10%)
Each
Quantity:
5 L
Request bulk or custom format
Price (USD)
402.65
Online Exclusive
447.00
Save 44.35 (10%)
Each
Specifications
Chemical Name or MaterialTBS with Tween (TBST)
Recommended StorageAmbient temperatures
Concentration20X
Physical FormSolution
Quantity5 L
View more
Used in Western blotting protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only