Sodium Dodecylsulfate, 10% Solution, Ultrapure
Sodium Dodecylsulfate, 10% Solution, Ultrapure
Sodium Dodecylsulfate, 10% Solution, Ultrapure
Thermo Scientific Chemicals

Sodium Dodecylsulfate, 10% Solution, Ultrapure

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
Catalog number J77504.AE
Price (USD)
74.10
Each
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Price (USD)
74.10
Each
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Absorbance (280 nm, 3%, 1 cm):≤ 0.1
Assay (unspecified)Absorbance (230 nm, 3%, 1 cm):≤ 0.2
RUO – Research Use Only