Owl™ JGC Gel Casting Systems
Thermo Scientific™

Owl™ JGC Gel Casting Systems

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
Have Questions?
Catalog NumberLength (Metric) Gel
JGC-214 cm
Catalog number JGC-2
Price (USD)
234.46
Online Exclusive
486.00
Save 251.54 (52%)
Each
Length (Metric) Gel:
14 cm
Price (USD)
234.46
Online Exclusive
486.00
Save 251.54 (52%)
Each

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

  • Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
  • Design allows storage of gels without dehydration by adding buffer and heat sealing.
  • System casts from one to four gels.
  • Few moving parts result in outstanding durability, extra long life.

Includes:

  • Casting stand
  • Blocking plate
  • Three-point leveling base
  • Spacer placer
  • 25 gel pouches
* Gel plates and combs sold separately
Specifications
Depth (English)6.69 in.
Depth (Metric)17 cm
Length (Metric) Gel14 cm
Width (English)7.06 in.
Width (Metric)18 cm
Width (Metric) Gel16 cm
For Use With (Application)Gel Electrophoresis
For Use With (Equipment)Owl JGC Gel Casting System
Gel Size20 x 20 cm
Product LineOwl
TypeGel Caster
Unit SizeEach