Sensititre™ Breakpoint Eye Two-Isolate Plate
Sensititre™ Breakpoint Eye Two-Isolate Plate
Sensititre™ Breakpoint Eye Two-Isolate Plate
Thermo Scientific™
Sensititre™ Breakpoint Eye Two-Isolate Plate
Perform accurate AST with Thermo Scientific™ Sensititre™ Breakpoint Eye Two-Isolate Plate, a two-isolate, 96-well microtitre plate for breakpoint testing of topical compounds.
Have Questions?
Catalog NumberQuantity
JOEYE210/Pk.
Catalog number JOEYE2
Price (ZAR)
-
Quantity:
10/Pk.
Perform accurate AST with the Thermo Scientific™ Sensititre™ Breakpoint Eye Two-Isolate Plate, a two-isolate, 96-well microtitre plate for breakpoint testing of topical compounds.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionBreakpoint Eye Two-Isolate Plate
Isolates Per Plate2
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Reading OptionsManual
Specimen TypeTwo-Isolate
Product TypeAST Plate
SpeciesAll
Target Organism ClassNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test TypeBreakpoint
Unit SizeEach