Thermolyne™ Premium Large Muffle Furnaces Accessories
Thermo Scientific™

Thermolyne™ Premium Large Muffle Furnaces Accessories

Use these accessories with Thermolyne Premium Large Muffle Furnaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
JSX16Shelf Pegs
PH177X1Hearth Tray, Large
PHX1Hearth Tray, Small
SH408X1Shelf
Catalog number JSX16
Price (USD)
-
Type:
Shelf Pegs

Use Thermo Scientific™ Thermolyne Premium Large Muffle Furnace Accessories to increase the utility of the Large Muffle Furnace.

Specifications
DescriptionShelf Pegs
For Use With (Equipment)Thermolyne Premium Large Muffle Furnace
Temperature (Metric)100°C to 1200°C
TypeShelf Pegs
Voltage0/1 V
Unit SizeEach