Long PCR Enzyme Mix (5 U/µL) - FAQs
Long PCR Enzyme Mix (5 U/µL) - FAQs
View additional product information for Long PCR Enzyme Mix (5 U/µL) - FAQs (K0181)
1 product FAQs found
How important is the quality of my DNA template in long PCR?
Template preparation becomes particularly important when performing longer amplifications (>15 kb). Therefore, it is recommended to check the length of the DNA by agarose gel electrophoresis.