IMPORTANT INFORMATION: The correct speed and time for high-speed centrifugation is 48,000 × g for 20 minutes, instead of 4,000–5,000 × g for 40 minutes, as indicated on page 13, step 4 of the instructions for purifying plasmid DNA using high-speed centrifugation.
Product Image
Thermo Scientific™

GeneJET Plasmid Midiprep Kit

Designed for large-scale isolation of high quality plasmid DNA from recombinant E.coli cultures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K0482100 Preps
K048125 Preps
Catalog number K0482
Price (USD)
911.65
Online exclusive
1,060.00
Save 148.35
Each
Quantity:
100 Preps
Price (USD)
911.65
Online exclusive
1,060.00
Save 148.35
Each

Thermo Scientific GeneJET Plasmid Midiprep Kit, sufficient for 25 preps, is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. The kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 200 μg of high copy number plasmid DNA. The purified plasmid DNA can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient—high yields up to 200 μg of high quality plasmid DNA from 50 mL of bacterial culture
  • High concentration of DNA purified— >400 ng/μL
  • Convenient—spin column format, centrifuge or vacuum protocols
  • Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns

Applications

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription
  • Transfection of robust cell lines
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeSpin Column
Concentration400 ng/μL
For Use With (Application)Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Prep Scale100-200 μg (Medium-Scale) Plasmid DNA
Product LineGeneJET
Product TypePlasmid MidiPrep Kit
Quantity100 Preps
Sample TypeBacteria
ScaleMidi
TargetPlasmid DNA
Test Time60 min.
FormatSpin Column
Isolation TechnologySilica Column
Unit SizeEach

Frequently asked questions (FAQs)

Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET plasmid purification kits compatible with EndA+ bacterial strains?

Yes, MagJET and GeneJET Plasmid DNA Purification Kits are suitable for plasmid DNA purification from EndA+ strains. Please refer to the specific kit protocol for details. GeneJET Plasmid Miniprep Kit requires Wash Solution I (Cat. No. R1611, not included in the kit) for EndA+ strains.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

Citations & References (1)

Citations & References
Abstract
Most Commonly Mutated Genes in High-Grade Serous Ovarian Carcinoma Are Nonessential for Ovarian Surface Epithelial Stem Cell Transformation.
Authors:Yamulla RJ,Nalubola S,Flesken-Nikitin A,Nikitin AY,Schimenti JC
Journal:Cell reports
PubMed ID:32877668
High-grade serous ovarian carcinoma (HGSOC) is the fifth leading cause of cancer-related deaths of women in the United States. Disease-associated mutations have been identified by the Cancer Genome Atlas Research Network. However, aside from mutations in TP53 or the RB1 pathway that are common in HGSOC, the contributions of mutation ... More