We have updated the storage temperature of the purification columns in this product. For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
GeneJET Plant RNA Purification Kit
Thermo Scientific™

GeneJET Plant RNA Purification Kit

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K0802250 Preps
K080150 Preps
Catalog number K0802
Price (USD)
1,000.65
Online exclusive
1,266.00
Save 265.35
Each
Quantity:
250 Preps
Price (USD)
1,000.65
Online exclusive
1,266.00
Save 265.35
Each

Thermo Scientific GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Highlights

  • Fast—procedure completed in 20 min following cell lysis
  • Convinient—protocol does not include clogging-prone lysate filtration step
  • High yield—up to 65 μg of high purity and high integrity RNA from 50 mg plant sample
  • Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)

Applications

Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:

  • RT-PCR
  • RT-qPCR
  • Northern blotting
  • Nuclease protection assay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume50 μL
Final Product TypeTotal RNA
For Use With (Application)RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Purification Time20 min. (After sample homogenization)
Quantity250 Preps
ScaleMini
Shipping ConditionRoom Temperature
Starting Material Amount≤100 mg
YieldUp to 65 μg
Isolation TechnologySilica Spin Column
Sample TypePlants
Unit SizeEach
Contents & Storage
• 200 mL Plant RNA Lysis Solution; store at room temperature
• 200 mL Wash Buffer WB 1 (concentrated); room temperature
• 2 x 100 mL Wash Buffer 2 (concentrated); room temperature
• 125 mL Water, Nuclease-free; room temperature
• 250 GeneJET RNA Spin Columns with Collection Tubes; For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.
• 250 Collection Tubes (2 mL); room temperature
• 250 Collection Tubes (1.5 mL); room temperature

Frequently asked questions (FAQs)

Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Is any GeneJET kit suitable for RNA purification from formalin-fixed paraffin-embedded FFPE tissues?

We cannot recommend GeneJET kits for RNA purification from FFPE tissues. In case of genomic DNA purififcation from formalin-fixed paraffin-embedded samples, we recommend GeneJET FFPE DNA Purification Kit.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.