GeneJET RNA Cleanup and Concentration Micro Kit
Actual product may vary
Thermo Scientific™

GeneJET RNA Cleanup and Concentration Micro Kit

GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K0842250 Preps
K084150 Preps
Catalog number K0842
Price (USD)
740.65
Online exclusive
832.00
Save 91.35
Each
Quantity:
250 Preps
Price (USD)
740.65
Online exclusive
832.00
Save 91.35
Each

GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Thermo Scientific GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Highlights

  • Advanced column design—easy to open, load and handle.
  • Elute in 6 to 10 μL volume.
  • 85 to 100% recovery of a high integrity RNA.
  • Fast and simple—procedure takes less than 5 minutes.

Applications:

  • The purified RNA can be used in common downstream applications
  • RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions250 Preps
Product LineGeneJET
Purification Time5 min.
Quantity250 Preps
ScaleMicro
FormatSpin Column
Isolation TechnologySpin Column
Sample TypeRNA
TargetTotal RNA
Unit SizeEach
Contents & Storage
For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.

Frequently asked questions (FAQs)

Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Is any GeneJET kit suitable for RNA purification from formalin-fixed paraffin-embedded FFPE tissues?

We cannot recommend GeneJET kits for RNA purification from FFPE tissues. In case of genomic DNA purififcation from formalin-fixed paraffin-embedded samples, we recommend GeneJET FFPE DNA Purification Kit.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.